Learn More
Novus Biologicals™ Human Human IgG, Fab Protein
Human Isotype Control Antibody
Manufacturer:  Novus Biologicals™ NBP197020
Catalog No. NBP197020
Add to cart
Description
Description
IgG F(ab) Isotype Control antibody specifically detects IgG F(ab) in Human samples. It is validated for Western Blot, ELISA, SDS-Page.
Specifications
Specifications
WB, ELISA, IP
Human IgG, Fab Protein
Human
Human
0.01 M Sodium Phosphate, 0.15M Sodium Chloride, pH 7.2.
4°C short term, −20°C long term. Avoid freeze-thaw cycles.
Human IgG Fab fragment assayed by immunoelectrophoresis resulted in a single precipitin arc against anti-Human Serum, anti- Human IgG and anti- Human IgG F(ab)2. No reaction was observed against anti- Human IgG F(c) or anti-Papain.