Learn More
Novus Biologicals™ Human Human IgG, Fc Protein
Human Isotype Control Antibody
Manufacturer:  Novus Biologicals™ NBP196973
Catalog No. NBP196973
Add to cart
Description
Description
IgG Fc Isotype Control antibody specifically detects IgG Fc in Human samples. It is validated for Western Blot, ELISA, Dot Blot, SDS-Page.
Specifications
Specifications
Western Blot, ELISA, Dot Blot, SDS-Page
0.02 M Potassium Phosphate, 0.15M Sodium Chloride, pH 7.2
Western Blot 1:1000, ELISA 1:5000-1:50000, Dot Blot, SDS-Page
Human
Human
ELISA, IP
Human IgG, Fc Protein
4°C short term, −20°C long term. Avoid freeze-thaw cycles.
Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Human Serum, anti-Human IgG and anti-Human IgG F(c). No reaction was observed against anti-Human IgG F(ab)2 or anti-Papain.