Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Recombinant Human Josephin1 His Protein
Highly purified. Generating reliable and reproducible results.
Supplier: Novus Biologicals™ NBP223156
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-202 of Human Josephin1 The Recombinant Human Josephin1 Protein is derived from E. coli. The Recombinant Human Josephin1 Protein has been validated for the following applications: SDS-Page.Specifications
NP_055691 | |
20mM Tris-HCl buffer (pH 8.0), 0.2 M NaCl, 40% glycerol, 5mM DTT, 2mM EDTA | |
25.6kDa | |
SDS-PAGE | |
E. Coli | |
−20°C or −80°C. Avoid freeze-thaw cycles. | |
Unconjugated | |
Recombinant |
SDS-PAGE | |
9929 | |
Josephin1 Protein | |
0.1mg | |
JOSD1, 1-202aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSMSCVPWK GDKAKSESLE LPQAAPPQIY HEKQRRELCA LHALNNVFQD SNAFTRDTLQ EIFQRLSPNT MVTPHKKSML GNGNYDVNVI MAALQTKGYE AVWWDKRRDV GVIALTNVMG FIMNLPSSLC WGPLKLPLKR QHWICVREVG GAYYNLDSKL KMPEWIGGES ELRKFLKHHL RGKNCELLLV VPEEVEAHQS WRTDV | |
RUO | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction