Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Recombinant Human KISS1 Protein
Highly purified. Generating reliable and reproducible results.
Supplier: Novus Biologicals™ NBC118516
Description
An un-tagged recombinant protein corresponding to the amino acids 1-120 of Human KISS1 The Recombinant Human KISS1 Protein is derived from E. coli. The Recombinant Human KISS1 Protein has been validated for the following applications: SDS-Page.Specifications
Q15726 | |
SDS-PAGE | |
3814 | |
KISS1 Protein | |
0.1 mg | |
KISS1, 1-120aa, Human, Recombinant, E.coli (NP_002247). | |
RUO | |
Unconjugated | |
Recombinant | |
>90%, by SDS-PAGE |
1 mg/mL | |
20 mM Tris-HCl buffer (pH 8.0), 1 mM DTT, 10% glycerol | |
M.W. (theoretical): 12.6 kDa | |
Protein | |
E. Coli | |
Store at 4 C short term. Aliquot and store (at -20 C long term. Avoid freeze-thaw cycles.) | |
KISS1 | |
Human | |
Cancer | |
KISS1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction