Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human Lactate Dehydrogenase A/LDHA Recombinant Protein

Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Functional, SDS-Page, Bioactivity

Manufacturer:  Novus Biologicals™ NBP140407

Catalog No. NBP140407


Add to cart

Description

Description

A bioactive recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-332 of Human Lactate Dehydrogenase A/LDHA The Recombinant Human Lactate Dehydrogenase A/LDHA Protein is derived from E. coli. The Recombinant Human Lactate Dehydrogenase A/LDHA Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.
Specifications

Specifications

NP_005557
20mM Tris-HCl buffer (pH8.0), 20% glycerol, 0.1 M NaCl
38.8kDa
Protein
0.1mg
1-332 aa Human, His-tagged, MGSSHHHHHH SSGLVPRGSH MATLKDQLIY NLLKEEQTPQ NKITVVGVGA VGMACAISIL MKDLADELAL VDVIEDKLKG EMMDLQHGSL FLRTPKIVSG KDYNVTANSK LVIITAGARQ QEGESRLNLV QRNVNIFKFI IPNVVKYSPN CKLLIVSNPV DILTYVAWKI SGFPKNRVIG SGCNLDSARF RYLMGERLGV HPLSCHGWVL GEHGDSSVPV WSGMNVAGVS LKTLHPDLGT DKDKEQWKEV HKQVVESAYE VIKLKGYTSW AIGLSVADLA ESIMKNLRRV HPVSTMIKGL YGIKDDVFLS VPCILGQNGI SDLVKVTLTS EEEARLKKSA DTLWGIQKEL QF
RUO
Human
ELISA, SDS-Page
3939
Lactate Dehydrogenase A/LDHA Protein
>95%
E. Coli
−80°C. Avoid freeze-thaw cycles.
Unlabeled
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit