Learn More
Novus Biologicals™ Human Lymphotoxin-alpha/TNF-beta Protein
Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Functional, SDS-Page, Bioactivity
Manufacturer:  Novus Biologicals™ NBP141147
Catalog No. NBP141147
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
A bioactive recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 35-205 of Human Lymphotoxin-alpha/TNF-beta The Recombinant Human Lymphotoxin-alpha/TNF-beta Protein is derived from E. coli. The Recombinant Human Lymphotoxin-alpha/TNF-beta Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.
Specifications
Specifications
NP_001153212
20mM Tris-HCl buffer (pH8.0), 30% glycerol, 0.1 M NaCl, 1mM DTT
20.9kDa
Protein
MGSSHHHHHH SSGLVPRGSH MLPGVGLTPS AAQTARQHPK MHLAHSTLKP AAHLIGDPSK QNSLLWRANT DRAFLQDGFS LSNNSLLVPT SGIYFVYSQV VFSGKAYSPK ATSSPLYLAH EVQLFSSQYP FHVPLLSSQK MVYPGLQEPW LHSMYHGAAF QLTQGDQLST HTDGIPHLVL SPSTVFFGAF AL
Human
ELISA, SDS-Page
4049
Lymphotoxin-alpha/TNF-beta Protein
>90%
−80°C. Avoid freeze-thaw cycles.