Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human Nanog Recombinant Protein

Highly purified. Generating reliable and reproducible results.

Manufacturer:  Novus Biologicals™ NBP142463

Catalog No. NBP142463


Add to cart

Description

Description

Recombinant Nanog protein with a His tag corresponding to amino acids 1-305. Source: E. coli (Accession #: Q9H9S0) The Recombinant Human Nanog His Protein is derived from E. coli. The Recombinant Human Nanog His Protein has been validated for the following applications: Western Blot, SDS-Page.
Specifications

Specifications

Q9H9S0
50mM Tris pH 8.0, 350mM NaCl, 8 M Urea, 50mM Imidazole
Nanog Protein
>95%
E. Coli
−80°C. Avoid freeze-thaw cycles.
Unlabeled
Yes
WB, SDS-Page
79923
Protein
0.01mg
Full length Homeobox protein NANOG derived from Human. The protein is His-tagged and recombinantly expressed in E. coli.
RUO
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit