Learn More
Novus Biologicals™ Human NQO-1 Recombinant Protein
Highly purified. Generating reliable and reproducible results.
Manufacturer:  Novus Biologicals™ NBP130240
Catalog No. NBP130240
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-274 of Human NQO-1 The Recombinant Human NQO-1 Protein is derived from E. coli. The Recombinant Human NQO-1 Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
P15559
20mM Tris-HCl buffer (pH 8.0), 10% glycerol, 1mM DTT
33kDa
Protein
0.1mg
NQO1, 1-274 aa, Human, His-tagged, Recombinant, E.coli (NP_000894)
RUO
Human
ELISA, SDS-Page
1728
NQO-1 Protein
>95%
E. Coli
−80°C. Avoid freeze-thaw cycles.
Unlabeled
Yes