Learn More
Novus Biologicals™ Human p23/PTGES3 Protein
Highly purified. Generating reliable and reproducible results.
Manufacturer:  Novus Biologicals™ NBP130281
Catalog No. NBP130281
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
An un-tagged recombinant protein corresponding to the amino acids 1-160 of Human p23/PTGES3 The Recombinant Human p23/PTGES3 Protein is derived from E. coli. The Recombinant Human p23/PTGES3 Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
NP_006592
20mM Tris-HCl buffer (pH8.0), 1mM DTT, 10% glycerol
18.6kDa
Protein
MQPASAKWYD RRDYVFIEFC VEDSKDVNVN FEKSKLTFSC LGGSDNFKHL NEIDLFHCID PNDSKHKRTD RSILCCLRKG ESGQSWPRLT KERAKLNWLS VDFNNWKDWE DDSDEDMSNF DRFSEMMNNM GGDEDVDLPE VDGADDDSQD SDDEKMPDLE
Human
ELISA, SDS-Page
10728
p23/PTGES3 Protein
>95%
−80°C. Avoid freeze-thaw cycles.