Learn More
Novus Biologicals™ Human PNMT Protein
Highly purified. Generating reliable and reproducible results.
Manufacturer:  Novus Biologicals™ NBP130280
Catalog No. NBP130280
Add to cart
Description
Description
An un-tagged recombinant protein corresponding to the amino acids 1-282 of Human PNMT The Recombinant Human PNMT Protein is derived from E. coli. The Recombinant Human PNMT Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
NP_002677
20mM Tris-HCl buffer (pH8.0), 10% glycerol
30.8kDa
Protein
MSGADRSPNA GAAPDSAPGQ AAVASAYQRF EPRAYLRNNY APPRGDLCNP NGVGPWKLRC LAQTFATGEV SGRTLIDIGS GPTVYQLLSA CSHFEDITMT DFLEVNRQEL GRWLQEEPGA FNWSMYSQHA CLIEGKGECW QDKERQLRAR VKRVLPIDVH QPQPLGAGSP APLPADALVS AFCLEAVSPD LASFQRALDH ITTLLRPGGH LLLIGALEES WYLAGEARLT VVPVSEEEVR EALVRSGYKV RDLRTYIMPA HLQTGVDDVK GVFFAWAQKV GL
Human
ELISA, SDS-Page
5409
PNMT Protein
>95%
−80°C. Avoid freeze-thaw cycles.