Learn More
Novus Biologicals™ Human Profilin 4 Protein
Highly purified. Generating reliable and reproducible results.
Manufacturer:  Novus Biologicals™ NBP141134
Catalog No. NBP141134
Add to cart
Description
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-129 of Human Profilin 4 The Recombinant Human Profilin 4 Protein is derived from E. coli. The Recombinant Human Profilin 4 Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
NP_955378
20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 10% glycerol
16.4kDa
Protein
MGSSHHHHHH SSGLVPRGSH MSHLQSLLLD TLLGTKHVDS AALIKIQERS LCVASPGFNV TPSDVRTLVN GFAKNPLQAR REGLYFKGKDYRCVRADEYS LYAKNENTGV VVVKTHLYLL VATYTEGMYP SICVEATESL GDYLRKKGS
Human
ELISA, SDS-Page
375189
Profilin 4 Protein
>95%
−80°C. Avoid freeze-thaw cycles.