Learn More
Novus Biologicals™ Human RAC2 Protein
Highly purified. Generating reliable and reproducible results.
Manufacturer:  Novus Biologicals™ NBP130199
Catalog No. NBP130199
Add to cart
Description
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-189 of Human RAC2 The Recombinant Human RAC2 Protein is derived from E. coli. The Recombinant Human RAC2 Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
NP_002863
20mM Tris-HCl buffer (pH 8.0), 20% glycerol, 2mM EDTA, 2mM DTT, 0.1 M NaCl
23.3kDa
Protein
MGSSHHHHHH SSGLVPRGSH MQAIKCVVVG DGAVGKTCLL ISYTTNAFPG EYIPTVFDNY SANVMVDSKP VNLGLWDTAG QEDYDRLRPL SYPQTDVFLI CFSLVSPASY ENVRAKWFPE VRHHCPSTPI ILVGTKLDLR DDKDTIEKLK EKKLAPITYP QGLALAKEID SVKYLECSAL TQRGLKTVFD EAIRAVLCPQ PTRQQKRAC
Human
ELISA, SDS-Page
5880
RAC2 Protein
>95%
−80°C. Avoid freeze-thaw cycles.