Learn More
Novus Biologicals™ Human Serpin A3/alpha 1-Antichymotrypsin Recombinant Protein
Highly purified. Generating reliable and reproducible results.
Manufacturer:  Novus Biologicals™ NBP130283
Catalog No. NBP130283
Add to cart
Description
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 24-423 of Human Serpin A3/alpha 1-Antichymotrypsin The Recombinant Human Serpin A3/alpha 1-Antichymotrypsin Protein is derived from E. coli. The Recombinant Human Serpin A3/alpha 1-Antichymotrypsin Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
P01011
20mM Tris-HCl buffer (pH 8.0), 10% glycerol, 1mM DTT
47.6kDa
Protein
0.1mg
Alpha-1-antichymotrypsin, 24-423 aa, Human, His-tag, E.coli (NP_001076).
RUO
Human
ELISA, SDS-Page
12
Serpin A3/alpha 1-Antichymotrypsin Protein
>90%
E. Coli
−80°C. Avoid freeze-thaw cycles.
Unlabeled
Yes