Learn More
Novus Biologicals™ Human Serpin A5/Protein C Inhibitor Protein
Highly purified. Generating reliable and reproducible results.
Manufacturer:  Novus Biologicals™ NBP130282
Catalog No. NBP130282
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 20-406 of Human Serpin A5/Protein C Inhibitor The Recombinant Human Serpin A5/Protein C Inhibitor Protein is derived from E. coli. The Recombinant Human Serpin A5/Protein C Inhibitor Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
AAH08915
20mM MES buffer (pH6.0), 150mM NaCl, 10% glycerol
45.9kDa
Protein
MGSSHHHHHH SSGLVPRGSH MHRHHPREMK KRVEDLHVGA TVAPSSRRDF TFDLYRALAS AAPSQNIFFS PVSISMSLAM LSLGAGSSTK MQILEGLGLN LQKSSEKELH RGFQQLLQEL NQPRDGFQLS LGNALFTDLV VDLQDTFVSA MKTLYLADTF PTNFRDSAGA MKQINDYVAK QTKGKIVDLL KNLDSNAVVI MVNYIFFKAK WETSFNHKGT QEQDFYVTSE TVVRVPMMSR EDQYHYLLDR NLSCRVVGVP YQGNATALFI LPSEGKMQQV ENGLSEKTLR WLKMFKKRQ LELYLPKFSI EGSYQLEKVL PSLGISNVFT SHADLSGISN HSNIQVSEMV HKAVVEVDES GTRAAAATGT IFTFRSARLN SQRLVFNRPF LMFIVDNNIL FLGKVNRP
Human
ELISA, SDS-Page
5104
Serpin A5/Protein C Inhibitor Protein
>90%
−80°C. Avoid freeze-thaw cycles.