Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human Serum Amyloid A1 Recombinant Protein

Manufacturer:  Novus Biologicals™ NBP130209SS

Catalog No. NBP130209SS

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The purity of this protein is >95% by SDS-PAGE. Molecular weight is 13.9kDa (125aa), confirmed by MALDI-TOF.
Specifications

Specifications

P02735
In 20mM Tris buffer(pH 8.0) containing 10% glycerol.
13.9kDa
Protein
0.02mg
Serum amyloid A, 19-122 aa, Human, His-tagged, Recombinant, E.coli (AAI_05797).
RUO
Human
SDS-Page
6288
Serum Amyloid A1 Protein
>95%
E. Coli
−80°C. Avoid freeze-thaw cycles.
Unlabeled
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit