Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human Thymidylate Synthase Recombinant Protein

Highly purified. Generating reliable and reproducible results.

Manufacturer:  Novus Biologicals™ NBP130310

Catalog No. NBP130310


Add to cart

Description

Description

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-313 of Human Thymidylate Synthase The Recombinant Human Thymidylate Synthase Protein is derived from E. coli. The Recombinant Human Thymidylate Synthase Protein has been validated for the following applications: SDS-Page.
Specifications

Specifications

P04818
20mM Tris-HCl buffer (pH 8.0), 10% glycerol, 1mM DTT, 0.1 M NaCl
37.8kDa
Protein
0.1mg
Thymidylate synthase, 1-313aa, Human, His tag, E.coli (NP_001062).
RUO
Human
SDS-Page
7298
Thymidylate Synthase Protein
>95%
E. Coli
4°C short term, −20°C long term. Avoid freeze-thaw cycles.
Unlabeled
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit