Learn More
Novus Biologicals™ Human TSTD1 Protein
Highly purified. Generating reliable and reproducible results.
Manufacturer:  Novus Biologicals™ NBP17879850UG
Catalog No. NBP17879850
Add to cart
Description
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-115 of Human TSTD1 The Recombinant Human TSTD1 Protein is derived from E. coli. The Recombinant Human TSTD1 Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
Q8NFU3
Liquid. In 20mM Tris-HCl buffer (pH 8.0) containing 1mM DTT, 10% glycerol,100mM NaCl
14.6kDa
SDS-PAGE
4°C short term, −20°C long term. Avoid freeze-thaw cycles.
SDS-Page
100131187
TSTD1 Protein
>95%
Human