Learn More
Novus Biologicals™ Human UBE2S Recombinant Protein
Highly purified. Generating reliable and reproducible results.
Manufacturer:  Novus Biologicals™ NBP130208
Catalog No. NBP130208
Add to cart
Description
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-222 of Human UBE2S The Recombinant Human UBE2S Protein is derived from E. coli. The Recombinant Human UBE2S Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
Q16763
20mM Tris-HCl buffer (pH8.0), 1mM DTT, and 20% glycerol
27.9kDa
Protein
0.1mg
UBE2S, 1-222 aa, Human, His-tagged, Recombinant, E.coli (NP_055316.2).
RUO
Human
ELISA, SDS-Page
27338
UBE2S Protein
>90%
E. Coli
−80°C. Avoid freeze-thaw cycles.
Unlabeled
Yes