Learn More
Novus Biologicals™ Human Ubiquitin Protein
Highly purified. Generating reliable and reproducible results.
Manufacturer:  Novus Biologicals™ NBP142465
Catalog No. NBP142465
Description
Description
Recombinant ubiquitin protein with an N-terminal His tag and corresponding to amino acids 1-156. Source: E. coli (Accession #: P62979) The Recombinant Human Ubiquitin His Protein is derived from E. coli. The Recombinant Human Ubiquitin His Protein has been validated for the following applications: Western Blot, SDS-Page.
Specifications
Specifications
P62979
25mM Tris pH 8.0, 250mM NaCl, 8 M Urea, 1mM DTT
Ubiquitin Protein
−80°C. Avoid freeze-thaw cycles.
WB, SDS-Page
6233
SDS-PAGE
Human