Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Human VEGF 121 Recombinant Protein
Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Functional, SDS-Page, Bioactivity
Supplier: Novus Biologicals™ NBC11852050UG
Description
A bioactive recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 207-327 of Human VEGF 121 The Recombinant Human VEGF 121 Protein is derived from E. coli. The Recombinant Human VEGF 121 Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.Specifications
P15692 | |
Liquid in 20mM Tris pH 8.0 containing 10% glycerol | |
16.3kDa | |
Protein | |
E. Coli | |
−80°C. Avoid freeze-thaw cycles. | |
Unconjugated | |
Recombinant |
ELISA, SDS-PAGE | |
7422 | |
VEGF 121 Protein | |
50μg | |
Amino acid 207-327 Isoform L-VEGF121, Human, His tag, E.coli (NP_001020541). | |
RUO | |
Human | |
>95% |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction