Learn More
Novus Biologicals™ Human Wnt-1 Protein
Highly purified and high bioactivity. Generating reliable and reproducible results.
Supplier: Novus Biologicals™ NBP199372
Description
Recombinant bioactive protein containing 343 amino acids for Human Wnt-1 The Recombinant Human Wnt-1 Protein is derived from E. coli. The Recombinant Human Wnt-1 Protein has been validated for the following applications: SDS-Page, Bioactivity.Specifications
P04628 | |
7471 | |
SDS-PAGE | |
−20°C. Avoid freeze-thaw cycles. |
Sterile filtered and lyophilized with no additives. | |
Wnt-1 Protein | |
>98%, by SDS-PAGE | |
Human |