Learn More
Novus Biologicals™ IKK alpha Peptide
Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
Manufacturer:  Novus Biologicals™ NBP176947PEP
Catalog No. NBP176947E
Add to cart
Description
Description
A blocking peptide containing 17 amino acids near the carboxy terminus of human IKK alpha (C2). Source: Synthetic (Accession #: AF009225) The IKK alpha Blocking Peptide has been validated for the following applications: Antibody Competition.
Specifications
Specifications
1147
For Blocking the binding of NBP1-76947
PBS pH 7.2 (10mM NAH2PO4, 10mM Na2HPO4, 130mM NaCl) containing 0.1% bovine serum albumin.
RUO
17 amino acids near the carboxy terminus of human IKK alpha (C2)
0.05mg
Synthetic
Human
−20°C. Avoid freeze-thaw cycles.
IKK alpha