Learn More
Novus Biologicals™ IKK beta Peptide
Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
Manufacturer:  Novus Biologicals™ NBP176935PEP
Catalog No. NBP176935E
Add to cart
Description
Description
A blocking peptide containing 19 amino acids near the carboxy terminus of human IKK beta (C3). Source: Synthetic (Accession #: O14920) The IKK beta Blocking Peptide has been validated for the following applications: Antibody Competition.
Specifications
Specifications
3551
For Blocking the binding of NBP1-76935
PBS pH 7.2 (10mM NAH2PO4, 10mM Na2HPO4, 130mM NaCl) containing 0.1% bovine serum albumin.
RUO
19 amino acids near the carboxy terminus of human IKK beta (C3)
0.05mg
Synthetic
Human
−20°C. Avoid freeze-thaw cycles.
IKK beta