Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ IL-12 Protein

Highly purified and high bioactivity. Generating reliable and reproducible results.

Manufacturer:  Novus Biologicals™ NBP1429160.5MG

 View more versions of this product

Catalog No. NBP1429165


Add to cart

Description

Description

Recombinant protein corresponding to amino acids Met1-Ser 328 of human IL-12. Accession #: NM_002187. Source: : Insect cells infected with baculovirus The Recombinant Human IL-12 Protein is derived from Sf 21 (baculovirus). The Recombinant Human IL-12 Protein has been validated for the following applications: ELISA, Immunoassay, Bioactivity.
Specifications

Specifications

ELISA, Immunoassay
0.5mg
IL-12
RUO
Unlabeled
Sterile liquid; 20mM NaH2PO4, pH 6.0, 0.2M NaCl, with no carriers or stabilizers. 0.22μm filtered.
Human
Store at -80°C. Avoid freeze-thaw cycles.
This recombinant human IL-12 p70 has been tested by sandwich ELISA using the human IL-12 p70 ELISA (B-T21/C8.6).
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit