Learn More
Novus Biologicals™ IL-27R alpha/WSX-1/TCCR Blocking Peptide
Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
Manufacturer:  Novus Biologicals™ NBP176727PEP
Catalog No. NBP176727E
Add to cart
Description
Description
A blocking peptide containing 15 amino acids near the carboxy terminus of human TCCR. Source: Synthetic The IL-27R alpha/WSX-1/TCCR Blocking Peptide has been validated for the following applications: Antibody Competition.
Specifications
Specifications
9466
For Blocking the binding of NBP1-76727
PBS pH 7.2 (10mM NAH2PO4, 10mM Na2HPO4, 130mM NaCl) containing 0.1% bovine serum albumin.
RUO
15 amino acids near the carboxy terminus of human TCCR
0.05mg
Synthetic
Human
−20°C. Avoid freeze-thaw cycles.
IL-27 R alpha/WSX-1/TCCR