Learn More
Novus Biologicals™ IL-3 Protein
Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Functional, SDS-Page, Bioactivity
Manufacturer:  Novus Biologicals™ NBP1511010.5MG
 View more versions of this product
Catalog No. NBP1511015
Add to cart
Description
Description
A recombinant protein with a His-tag corresponding to amino acids 20 to 152 of Human IL-3 The Recombinant Human IL-3 Protein is derived from E. coli. The Recombinant Human IL-3 Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.
Specifications
Specifications
1mg/mL
Liquid in 20mM Tris pH 8.0, 0.2mM PMSF, 10% glycerol
0.5mg
Store at -80°C. Avoid freeze-thaw cycles.
IL-3
SDS-Page
SDS-PAGE
Human
RUO
Unlabeled