Learn More
Novus Biologicals™ IL-4 Protein
Manufacturer:  Novus Biologicals™ NBP1429230.1MG
Catalog No. NBP14292301
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
ELISA, Immunoassay
0.1mg
IL-4
RUO
Unlabeled
Sterile liquid; phosphate buffered saline, pH 7.2, 150mM NaCl, with no carriers or preservatives. 0.22μm filtered.
Human
Store at -80°C. Avoid freeze-thaw cycles.
This recombinant human IL-4 has been tested by sandwich ELISA using anti-human IL-4 antibody pair (8D4-8/MP4-25D2).
Yes