Learn More
Novus Biologicals™ IL-6 Protein
Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Functional, SDS-Page, Bioactivity
Manufacturer:  Novus Biologicals™ NBP130184
Catalog No. NBP130184
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
A bioactive recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 30-212 of Human IL-6 The Recombinant Human IL-6 Protein is derived from E. coli. The Recombinant Human IL-6 Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.
Specifications
Specifications
NP_000591
PBS (pH 7.4), 10% glycerol
23.1kDa
Protein
MGSSHHHHHH SSGLVPRGSH MVPPGEDSKD VAAPHRQPLT SSERIDKQIR YILDGISALR KETCNKSNMC ESSKEALAEN NLNLPKMAEK DGCFQSGFNE ETCLVKIITG LLEFEVYLEY LQNRFESSEE QARAVQMSTK VLIQFLQKKA KNLDAITTPD PTTNASLLTK LQAQNQWLQD MTTHLILRSF KEFLQSSLRA LRQM
ELISA, SDS-Page
3569
IL-6 Protein
>95%
−80°C. Avoid freeze-thaw cycles.