Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Imiquimod, TLR7 Ligand

Applications: Functional, In vitro assay, Ligand Activation

$104.00

Specifications

For Use With (Application) For activation of TLR7
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Type Imiquimod, TLR7 Ligand
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP2262281
View Documents
Novus Biologicals™
NBP2262281MG
1mg
Each for $104.00
Add to cart
 
Description

Description

Imiquimod, TLR7 ligand is an imidazoquinoline amine analog to guanosine. As a synthetic molecule of the Imidazoquinoline family, it has potent immunostimulatory activity. Imiquimod has been shown to activate only TLR7, this activation is MyD88 dependant and leads to the induction of the transcription factor NF-kB.

  • Species: Human, Mouse
  • Specificity: Imiquimod is human/mouse TLR7 agonist
  • Stimulation of TLR7 can be achieved with 5-10μg/mL
Specifications

Specifications

For activation of TLR7
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Imiquimod, TLR7 Ligand
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit