Novus Biologicals™ Imm-Link™ BSA (Amine) Immunogen labeling Kit

Immunogen labeling kit

Manufacturer: Novus BiologicalsSupplier Diversity Partner 4520001

 View more versions of this product

Catalog No. 45-200-01

  Check Availability
Add to Cart

    Specifications

    Product Type Imm-Link BSA (Amine) Immunogen labeling Kit
    Quantity 1 x 2mg Reaction
    Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.

    BSA, Ovalbumin or Keyhole Limpet Hemocyanin (KLH) stimulate a large immune response and are typically used as carrier proteins to generate antibodies against small antigens (peptides or organic compounds). For success the antigen (normally referred to as a hapten) needs to be covalently linked to the carrier protein.

    The imm-Link™ amine conjugation kit allows amine containing haptens to be conjugated to a carrier protein simply by adding a solution of the hapten to a proprietary lyophilized mixture containing the carrier protein and all the required conjugation chemistry. The hands-on time to set up the conjugation reaction is typically 20-30 seconds.

    Following successful conjugation the conjugate is dialysed in the supplied dialysis cartridge to remove unwanted by products. The hapten conjugate can then be easily recovered and used.

    Order Info

    Quantity

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit