Learn More
Novus Biologicals™ Imm-Link™ BSA (Carboxyl) Immunogen labeling Kit
Immunogen labeling kit
Specifications
Product Type Imm-Link BSA (Carboxyl) Immunogen labeling Kit
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
47-200-01
View Documents
Novus Biologicals™
4720001
1 x 2mg Reaction This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
47-205-00
View Documents
Novus Biologicals™
4720500
3 x 2mg Reactions This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
BSA, Ovalbumin or Keyhole Limpet Hemocyanin (KLH) stimulate a large immune response and are typically used as carrier proteins to generate antibodies against small antigens (peptides or organic compounds). For success the antigen (normally referred to as a hapten) needs to be covalently linked to the carrier protein.
The imm-Link™ carboxyl conjugation kit allows carboxyl containing haptens to be conjugated to a carrier protein simply by adding a solution of the hapten to a proprietary lyophilised mixture containing the carrier protein and all the required conjugation chemistry. The hands-on time to set up the conjugation reaction is typically 20-30 seconds.
Following successful conjugation the conjugate is dialysed in the supplied dialysis cartridge to remove unwanted by products. The hapten conjugate can then be easily recovered and used.
Specifications
Specifications
Imm-Link BSA (Carboxyl) Immunogen labeling Kit