Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals Imm-Link BSA (Sulfhydryl) Immunogen labeling Kit
Immunogen labeling kit
Manufacturer: Novus Biologicals 4620001
Description
BSA, Ovalbumin or Keyhole Limpet Hemocyanin (KLH) stimulate a large immune response and are typically used as carrier proteins to generate antibodies against small antigens (peptides or organic compounds). For success the antigen (normally referred to as a hapten) needs to be covalently linked to the carrier protein.The imm-Link sulfhydryl conjugation kit allows sulfhydryl containing haptens to be conjugated to a carrier protein simply by adding a solution of the hapten to a proprietary lyophilised mixture containing the carrier protein and all the required conjugation chemistry. The hands-on time to set up the conjugation reaction is typically 20-30 seconds.
Following successful conjugation the conjugate is dialysed in the supplied dialysis cartridge to remove unwanted by products. The hapten conjugate can then be easily recovered and used.
Specifications
1 x 2mg Reaction | |
Store at -20°C. Avoid freeze-thaw cycles. | |
Imm-Link BSA (Sulfhydryl) Immunogen labeling Kit |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title