Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Imm-Link™ Ovalbumin (Amine) Immunogen labeling Kit

Immunogen labeling kit

Manufacturer:  Novus Biologicals™ 4510001

Catalog No. 45-100-01


Add to cart

Description

Description

BSA, Ovalbumin or Keyhole Limpet Hemocyanin (KLH) stimulate a large immune response and are typically used as carrier proteins to generate antibodies against small antigens (peptides or organic compounds). For success the antigen (normally referred to as a hapten) needs to be covalently linked to the carrier protein.

The imm-Link™ amine conjugation kit allows amine containing haptens to be conjugated to a carrier protein simply by adding a solution of the hapten to a proprietary lyophilised mixture containing the carrier protein and all the required conjugation chemistry. The hands-on time to set up the conjugation reaction is typically 20-30 seconds.

Following successful conjugation the conjugate is dialysed in the supplied dialysis cartridge to remove unwanted by products. The hapten conjugate can then be easily recovered and used.
Specifications

Specifications

1 x 2mg Reaction
Store at -20°C. Avoid freeze-thaw cycles.
Imm-Link Ovalbumin (Amine) Immunogen labeling Kit
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit