Learn More
Novus Biologicals™ InnovaCoat™ GOLD Antibody Labeling Kit, 40nm Mini
Antibody labeling kit
$320.85 - $688.85
Specifications
Product Type InnovaCoat GOLD 40nm Mini Antibody Labeling Kit
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
23-000-10
View Documents
Novus Biologicals™
2300010
10 Reactions Each for $688.85
Add to cart
23-000-05
View Documents
Novus Biologicals™
2300005
3 Reactions Each for $320.85
Add to cart
Description
Description
InnovaCoat nanoparticles in this kit are supplied as a freeze dried mixture. The conjugation reaction is initiated simply by adding a solution of the antibody, which becomes attached (via lysine residues) to the InnovaCoat surface. InnovaCoat GOLD can however be provided in a variety of formats (e.g. 10 OD liquid or as lyophilized nanoparticles) formulated to allow a one-step reaction with antibodies for bulk or custom orders. The kits are designed for research use and for the development of diagnostic products, specifically for the screening of potential antibodies or small scale proof-of-principle studies. The gold nanoparticles are also designed to be highly effective in lateral flow immunoassays.
Specifications
Specifications
InnovaCoat GOLD 40nm Mini Antibody Labeling Kit