Learn More
Novus Biologicals™ Integrin alpha 4/CD49d Peptide
Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
Manufacturer:  Novus Biologicals™ NBP176945PEP
Catalog No. NBP176945E
Add to cart
Description
Description
A blocking peptide containing 16 amino acids near the center of human integrin alpha 4. Source: Synthetic (Accession #: P13612) The Integrin alpha 4/CD49d Blocking Peptide has been validated for the following applications: Antibody Competition.
Specifications
Specifications
3676
For Blocking the binding of NBP1-76945
PBS pH 7.2 (10mM NAH2PO4, 10mM Na2HPO4, 130mM NaCl) containing 0.1% bovine serum albumin.
RUO
16 amino acids near the center of human integrin alpha 4#
0.05mg
Synthetic
Human
−20°C. Avoid freeze-thaw cycles.
Integrin alpha 4/CD49d