Learn More
Novus Biologicals™ JMJD6/PSR Peptide
Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
Manufacturer:  Novus Biologicals™ NBP177075PEP
Catalog No. NBP177075E
Add to cart
Description
Description
A blocking peptide containing 15 amino acids near the amino terminus of human JMJD6. Source: Synthetic (Accession #: Q6NYC1) The JMJD6/PSR Blocking Peptide has been validated for the following applications: Antibody Competition.
Specifications
Specifications
23210
For Blocking the binding of NBP1-77075
PBS pH 7.2 (10mM NAH2PO4, 10mM Na2HPO4, 130mM NaCl) containing 0.1% bovine serum albumin.
RUO
15 amino acids near the amino terminus of human JMJD6
0.05mg
Synthetic
Human
−20°C. Avoid freeze-thaw cycles.
JMJD6/PSR