Learn More
Novus Biologicals™ KCNE1 Overexpression Lysate (Native)
Manufacturer:  Novus Biologicals™ NBP209460
Catalog No. NBP209460
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
The KCNE1 Overexpression Lysate is derived from human.
Specifications
Specifications
The lysate was created in HEK293T cells, using plasmid ID RC215316 and based on accession number. The protein contains a C-terminal DDK tag.
RIPA buffer
Homo sapiens KCNE1, mRNA.
Native
0.1mg
Protein
-80°C. Avoid freeze-thaw cycles.
Western Blot