Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Lactate Dehydrogenase B Protein

Click to view available options
Quantity:
0.1 mg
0.5 mg
Description
A bioactive recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-334 of Human Lactate Dehydrogenase B The Recombinant Human Lactate Dehydrogenase B Protein is derived from E. coli. The Recombinant Human Lactate Dehydrogenase B Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.
Specifications
Specifications
Concentration | 1.0mg/mL |
For Use With (Application) | ELISA, SDS-PAGE |
Formulation | In 20mM Tris-HCl buffer (pH 8.0) containing 10% glycerol, 1mM DTT. |
Gene ID (Entrez) | 3945 |
Molecular Weight (g/mol) | 38.8kDa |
Purification Method | Protein |
Quantity | 0.5 mg |
Source | Human |
Immunogen | MGSSHHHHHH SSGLVPRGSH MATLKEKLIA PVAEEEATVP NNKITVVGVG QVGMACAISI LGKSLADELA LVDVLEDKLK GEMMDLQHGS LFLQTPKIVA DKDYSVTANS KIVVVTAGVR QQEGESRLNL VQRNVNVFKF IIPQIVKYSP DCIIIVVSNP VDILTYVTWK LSGLPKHRVI GSGCNLDSAR FRYLMAEKLG IHPSSCHGWI LGEHGDSSVA VWSGVNVAGV SLQELNPEMG TDNDSENWKE VHKMVVESAY EVIKLKGYTN WAIGLSVADL IESMLKNLSR IHPVSTMVKG MYGIENEVFL SLPCILNARG LTSVINQKLK DDEVAQLKKS ADTLWDIQKD LKDL |
Storage Requirements | Store at -80°C. Avoid freeze-thaw cycles. |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction