Learn More
Novus Biologicals™ LCE1C Overexpression Lysate
Supplier: Novus Biologicals™ NBP204976
Description
The LCE1C Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.Specifications
The lysate was created in HEK293T cells, using plasmid ID RC215316 and based on accession number NP_848128. The protein contains a C-terminal DDK tag. | |
0.1 mg | |
Protein | |
-80°C. Avoid freeze-thaw cycles. |
Western Blot | |
RIPA buffer | |
Homo sapiens LCE1C, mRNA. | |
Native |