Learn More
Novus Biologicals™ LDLR Blocking Peptide
Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
Manufacturer:  Novus Biologicals™ NBP177029PEP
Catalog No. NBP177029E
Add to cart
Description
Description
A blocking peptide containing 18 amino acids near the center of human LDL-R. Source: Synthetic (Accession #: NP_000518) The LDLR Blocking Peptide has been validated for the following applications: Antibody Competition.
Specifications
Specifications
3949
For Blocking the binding of NBP1-77029
PBS pH 7.2 (10mM NAH2PO4, 10mM Na2HPO4, 130mM NaCl) containing 0.1% bovine serum albumin.
RUO
18 amino acids near the center of human LDL-R
0.05mg
Synthetic
Human
−20°C. Avoid freeze-thaw cycles.
LDL R