Learn More
Novus Biologicals™ Lightning-Link™ Avidin Antibody Labeling Kit
Antibody labeling kit
Manufacturer:  Novus BiologicalsSupplier Diversity Partner 7170010
 View more versions of this product
Catalog No. 71-700-10
  Check Availability
Add to cart
Specifications
Description
Avidin is a 66kDa tetrameric glycoprotein composed of four identical subunits, which has widespread applications due to its very high affinity to the vitamin biotin.
Order Info
Quantity
Description & Specifications
Specifications
3 x 100μg Reactions
Store at -20°C. Avoid freeze-thaw cycles.
Avidin
Glass vial(s) of Lightning-Link mix (1 or 3 vials, depending on pack size), 1 vial of LL Modifier reagent, 1 vial of LL Quencher reagent
Avidin Antibody Labeling Kit