Novus Biologicals™ Lightning-Link™ Avidin Antibody Labeling Kit

Antibody labeling kit

Manufacturer: Novus BiologicalsSupplier Diversity Partner 7170015

 View more versions of this product

Catalog No. 71-700-15

  Check Availability
Add to Cart

    Specifications

    Product Type Avidin Antibody Labeling Kit
    Conjugate Avidin
    Quantity 1 x 1mg Reaction
    Kit Contents Glass vial(s) of Lightning-Link mix (1 or 3 vials, depending on pack size), 1 vial of LL Modifier reagent, 1 vial of LL Quencher reagent
    Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.

    Avidin is a 66kDa tetrameric glycoprotein composed of four identical subunits, which has widespread applications due to its very high affinity to the vitamin biotin.

    Order Info

    Quantity

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit