Novus Biologicals™ Lightning-Link™ Avidin Antibody Labeling Kit

Antibody labeling kit

Manufacturer: Novus BiologicalsSupplier Diversity Partner 7170030

 View more versions of this product

Catalog No. 71-700-30

  Check Availability
Add to Cart

    Specifications

    Product Type Avidin Antibody Labeling Kit
    Conjugate Avidin
    Quantity 3 x 0.01mg Reactions
    Kit Contents Glass vial(s) of Lightning-Link mix (1 or 3 vials, depending on pack size), 1 vial of LL Modifier reagent, 1 vial of LL Quencher reagent
    Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.

    Avidin is a 66kDa tetrameric glycoprotein composed of four identical subunits, which has widespread applications due to its very high affinity to the vitamin biotin. Activated avidin is covalently attached in a controlled process at near-neutral pH. The use of a stable avidin reagent at a low molar ratio of avidin:antibody, combined with highly efficient labeling process, means that desalting/dialysis steps are not needed and that avidin labeling can be carried out even at microgram scale without losses. The amount of antibody used for labeling should be the same as the pack size of LL-Avidin. For any new antibody, optimization of the ratio of antibody to dye is often worthwhile. If your antibody needs to be concentrated, purified, or cleaned up, consider purchasing an antibody clean up kit.

    Order Info

    Quantity

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit