Novus Biologicals™ Lightning-Link™ B-Phycoerythrin Antibody Labeling Kit

Antibody labeling kit

Manufacturer: Novus BiologicalsSupplier Diversity Partner 7160005

 View more versions of this product

Catalog No. 71-600-05

  Check Availability
Add to Cart

    Specifications

    Product Type B-Phycoerythrin Antibody Labeling Kit
    Conjugate B-Phycoerythrin
    Quantity 1 x 100μg Reaction
    Kit Contents Glass vial(s) of Lightning-Link mix (1 or 3 vials, depending on pack size), 1 vial of LL-B-PE Modifier reagent, 1 vial of LL-B-PE Quencher reagent
    Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.

    B-Phycoerythrin (B-PE) is a fluorescent protein from phycobiliprotein family. It has a primary absorption peak at 546 nm and a secondary peak at 563 nm. When excited at 546 nm it has a strong fluorescence emission with its maximum at 575 nm.

    Order Info

    Quantity

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit