Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Lightning-Link (R) Fluorescein Antibody Labeling Kit
Applications: Immunofluorescence
$542.50 - $1062.00
Specifications
Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
---|---|
Conjugate | FITC |
Kit Contents | Glass vial(s) of Lightning-Link mix (1 or 3 vials, depending on pack size), 1 vial of LL-AP Modifier reagent, 1 vial of LL-AP Quencher reagent |
Product Type | Fluorescein Antibody Labeling Kit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
70-700-30
|
Novus Biologicals™
7070030 |
30 μg |
Each for $542.50
|
|
|||||
70-700-10
|
Novus Biologicals™
7070010 |
300 μg |
Each for $808.00
|
|
|||||
70-700-15
|
Novus Biologicals™
7070015 |
1 mg |
Each for $1,062.00
|
|
|||||
Description
Fluorescein is a fluorophore widely used in biological analysis, with a pH dependent absorption and emission spectrum. In TBS LL-Fluorescein shows an absorption maximum at 494 nm and an emission maximum at 520 nm.Specifications
Store at -20°C. Avoid freeze-thaw cycles. | |
FITC | |
Glass vial(s) of Lightning-Link mix (1 or 3 vials, depending on pack size), 1 vial of LL-AP Modifier reagent, 1 vial of LL-AP Quencher reagent | |
Fluorescein Antibody Labeling Kit |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title