Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Lightning-Link (R) Glucose Oxidase Antibody Labeling Kit
Antibody labeling kit
$808.00 - $1183.00
Specifications
Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
---|---|
Conjugate | Gox |
Kit Contents | Glass vial(s) of Lightning-Link mix (1 or 3 vials, depending on pack size), 1 vial of LL-GOx Modifier reagent, 1 vial of LL-GOx Quencher reagent |
Product Type | Glucose Oxidase Antibody Labeling Kit |
Description
Glucose oxidase is an enzyme produced and purified from Aspergillus niger which catalyses the oxidation of glucose with the release of hydrogen peroxide. Its molecular weight is 80kDa, but it exists as a dimer of 160kDa Measurement of glucose by GOx is used in the food industry, in fermentation and, most importantly, as the basis of biosensors for the diagnosis of diabetes.Specifications
Store at -20°C. Avoid freeze-thaw cycles. | |
Gox | |
Glass vial(s) of Lightning-Link mix (1 or 3 vials, depending on pack size), 1 vial of LL-GOx Modifier reagent, 1 vial of LL-GOx Quencher reagent | |
Glucose Oxidase Antibody Labeling Kit |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title