Novus Biologicals™ Lightning-Link™ Glucose Oxidase Antibody Labeling Kit

Antibody labeling kit

Manufacturer: Novus BiologicalsSupplier Diversity Partner 7060015

 View more versions of this product

Catalog No. 70-600-15

  Check Availability
Add to Cart

    Specifications

    Product Type Glucose Oxidase Antibody Labeling Kit
    Conjugate Gox
    Quantity 1mg
    Kit Contents Glass vial(s) of Lightning-Link mix (1 or 3 vials, depending on pack size), 1 vial of LL-GOx Modifier reagent, 1 vial of LL-GOx Quencher reagent
    Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.

    Glucose oxidase is an enzyme produced and purified from Aspergillus niger which catalyses the oxidation of glucose with the release of hydrogen peroxide. Its molecular weight is 80kDa, but it exists as a dimer of 160kDa Measurement of glucose by GOx is used in the food industry, in fermentation and, most importantly, as the basis of biosensors for the diagnosis of diabetes.

    Order Info

    Quantity

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit