Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Lightning-Link (R) PE-Cy5 Antibody Labeling Kit
SDP

Applications: Flow Cytometry

Supplier:  Novus Biologicals™ 7600030

 View more versions of this product

Catalog No. 76-000-30


Add to Cart

Description

Description

PE/Cy5 is a tandem conjugate. The PE is excited at 488nm and functions as an energy donor for the Cy5. The energy is transferred via FRET (Fluorescence Resonance Energy Transfer) to the Cy5, which emits then the energy received in the form of long wavelength light at 670nm.

The antibody is covalently bonded to the label in a directional and controlled process at near-neutral pH. In view of the large size of the PE-Cy5 label, the amount of antibody used in a labeling reaction with the tandem must always be less than the pack size of LL PE-Cy5, in order that the PE-Cy5 is in a slight molar excess. The best ratio for any new antibody reagent must be determined by experimentation but 50-60ug of IgG antibody for every 100ug of LL PE-Cy5 usually gives optimal results. 50ug of antibody corresponds to an Ab:PE-Cy5 molar ratio of 1:1. The amount of antibody used for labeling should be the same as the pack size of LL-PE-Cy5. For any new antibody, optimization of the ratio of antibody to dye is often worthwhile. If your antibody needs to be concentrated, purified, or cleaned up, consider purchasing an antibody clean up kit.

Order Info

Quantity

Specifications

Specifications

30 μg
PE/Cy5
Flow Cytometry
Store at -20°C. Avoid freeze-thaw cycles.
Glass vial(s) of Lightning-Link mix (1 or 3 vials, depending on pack size), 1 vial of LL Modifier reagent, 1 vial of LL Quencher FD reagent
PE-Cy5 Antibody Labeling Kit
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit