Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Lightning-Link (R) R-PE Antibody Labeling Kit
Applications: Flow Cytometry
Supplier: Novus Biologicals™ 7030030
Description
R-Phycoerythrin (R-PE) is a fluorescent protein from the phycobiliprotein family, and is isolated from cyanobacteria, red algae and cryptomonads. The absorbance spectrum of R-PE covers a broad range of excitation wavelengths, which provides an advantage for multi-color immunofluorescent staining or cell sorting. R-PE is one of the most intensely fluorescent phycobiliprotein having orange fluorescence. It is significantly brighter and more photostable than conventional organic fluorophores and it has a high quantum yield. It has a strong absorption peak at 565 nm, and a secondary absorption peak at 492 nm. The emission peak is at 575. The amount of antibody used for labeling should be the same as the pack size of LL-R-PE. For any new antibody, optimization of the ratio of antibody to dye is often worthwhile. If your antibody needs to be concentrated, purified, or cleaned up, consider purchasing an antibody clean up kit.Order Info
Quantity
Specifications
30 μg | |
PE | |
R-PE Antibody Labeling Kit |
Store at -20°C. Avoid freeze-thaw cycles. | |
Glass vial(s) of Lightning-Link mix (1 or 3 vials, depending on pack size), 1 vial of LL-AP Modifier reagent, 1 vial of LL-AP Quencher reagent |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction