Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Lightning-Link (R) Rapid AMCA Antibody Labeling Kit
Antibody labeling kit
$542.50 - $1183.00
Specifications
Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
---|---|
Conjugate | AMCA |
Kit Contents | Glass vial(s) of Lightning-Link Rapid mix (1 or 3 vials, depending on pack size), 1 vial of Rapid Modifier reagent, 1 vial of Rapid Quencher reagent |
Product Type | Rapid AMCA Antibody Labeling Kit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
31-300-30
|
Novus Biologicals™
3130030 |
30 μg |
Each for $542.50
|
|
|||||
31-300-10
|
Novus Biologicals™
3130010 |
300 μg |
Each for $760.00
|
|
|||||
31-300-15
|
Novus Biologicals™
3130015 |
1 mg |
Each for $1,183.00
|
|
|||||
Description
Unlike standard AMCA labeling procedures, where a large molar excess of AMCA is employed, the Lightning-Link AMCA kit combines a low molar ratio of AMCA:antibody and high labeling efficiency to eliminate the need for purification steps. AMCA (aminomethylcoumarin acetate) is a blue fluorescent dye commonly used in fluorescence microscopy, arrays and in-situ hybridisation. It has an excitation wavelength of 345-350nm and an emission wavelength of 440-460nm.Specifications
Store at -20°C. Avoid freeze-thaw cycles. | |
AMCA | |
Glass vial(s) of Lightning-Link Rapid mix (1 or 3 vials, depending on pack size), 1 vial of Rapid Modifier reagent, 1 vial of Rapid Quencher reagent | |
Rapid AMCA Antibody Labeling Kit |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title